Pancreatin 8X, USP Grade
Product Image
MP Biomedicals
Pancreatin 8X, USP Grade
From porcine pancreas
Have Questions?
Catalog NumberQuantity
MPB021939762525 g
Catalog number MPB0219397625
Price (SGD)
87.00
EA
Add to cart
Quantity:
25 g
Price (SGD)
87.00
EA
Add to cart
Composed of numerous enzymes including amylase, trypsin, lipase, ribonuclease and protease. It will convert a minimum of 25 times its weight of potato starch into soluble carbohydrate in five minutes in water at 40°C, and not less than than 25 times its weight of casein into proteoses. It hydrolyzes fats to glycerol and fatty acid, protein to proteoses and derived compounds, and starch into dextrins and sugar. Lipase Activity: ≥16U/mg. Amylase Activity: ≥200U/mg. Protease Activity: ≥200U/mg.
Specifications
Concentration8X
ColorCream
For Use With (Application)Promote esophageal tissue loosening in patients with esophageal tumors
SourcePorcine pancreas
Quantity25 g
Product TypePancreatin
FormPowder
Unit SizeEA
Contents & Storage
0°C