Check Order Status
Ammonium Hydroxide, (ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Ammonium Hydroxide, (ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219385490
500 mL
Catalog number
MPB0219385490
Price (SGD)
111.00
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
111.00
Each
Add to cart
Specifications
Chemical Name or Material
Ammonium Hydroxide
CAS
1336-21-6
Recommended Storage
Room Temperature
Quantity
500 mL
Molecular Formula
H5NO
View more