Check Order Status
Calcium Acetate Monohydrate, ≥99%, (ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Calcium Acetate Monohydrate, ≥99%, (ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219382190
500 g
Catalog number
MPB0219382190
Price (SGD)
335.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
335.00
EA
Add to cart
Specifications
Chemical Name or Material
Calcium Acetate Monohydrate
CAS
5743-26-0
Recommended Storage
Room Temperature
Quantity
500 g
Assay Percent Range
≥99%
View more