Check Order Status
Calcium nitrate tetrhydrate, Reagent grade, For ACS analysis, ≥99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Calcium nitrate tetrhydrate, Reagent grade, For ACS analysis, ≥99%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219380183
250 g
Catalog number
MPB0219380183
Price (SGD)
86.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (SGD)
86.00
EA
Add to cart
Specifications
Chemical Name or Material
Calcium nitrate tetrhydrate
Name Note
For ACS analysis
CAS
13477-34-4
Color
White
Physical Form
Crystalline Powder
View more