▸
Biochemical Reagents
▸
Lipids and Lipid Derivatives
▸
Fatty acid conjugates
▸
Paclitaxel MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Paclitaxel MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219353201
1 mg
Catalog number
MPB0219353201
Price (SGD)
76.00
Each
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (SGD)
76.00
Each
Add to cart
Specifications
Chemical Name or Material
Paclitaxel
Name Note
From the Pacific Yew Tree
CAS
373-49-9
Color
White
Recommended Storage
Store at room temperature (15° to 30°C)
View more