4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride, MP Biomedicals
4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride, MP Biomedicals
4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride, MP Biomedicals
4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride, MP Biomedicals
MP Biomedicals
4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride, MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB0219350380100 mg
Catalog number MPB0219350380
Price (SGD)
273.00
EA
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (SGD)
273.00
EA
Add to cart
Specifications
Chemical Name or Material4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride
CAS30827-99-7
Molecular FormulaC8H11ClFNO2S
Quantity100 mg
Synonym4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
View more