Sodium Phosphate, Monobasic, Monohydrate, MP Biomedicals™
Sodium Phosphate, Monobasic, Monohydrate, MP Biomedicals™
Sodium Phosphate, Monobasic, Monohydrate, MP Biomedicals™
Sodium Phosphate, Monobasic, Monohydrate, MP Biomedicals™
MP Biomedicals
Sodium Phosphate, Monobasic, Monohydrate, MP Biomedicals™
Widely used in molecular biology, biochemistry and chromatography. Sodium Phosphate, Monobasic, Monohydrate, MP Biomedicals is a reagent with very high buffering capacity. It is is useful in conjunction with Sodium Phosphate, Dibasic in the preparation of
Have Questions?
Catalog NumberQuantity
MPB02191442.5500 g
Catalog number MPB02191442.5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
CAS10049-21-5
Recommended StorageRoom Temperature (15–30°C), desiccate.
Physical FormPowder
Quantity500 g
Molecular FormulaH5NaO5P
View more
Sodium phosphate occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4). Most neutral sodium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH for the application. IIt is used in many applications, including the purification of antibodies.