Potassium Phosphate Dibasic, MP Biomedicals™
Potassium Phosphate Dibasic, MP Biomedicals™
Potassium Phosphate Dibasic, MP Biomedicals™
Potassium Phosphate Dibasic, MP Biomedicals™
MP Biomedicals
Potassium Phosphate Dibasic, MP Biomedicals™
Used as a component for a variety of media used in the culture of microorganisms and in phosphate buffered saline (PBS). Potassium Phosphate Dibasic, MP Biomedicals is a reagent with high buffering capacity. In addition to helping maintain pH, it supplies
Have Questions?
Catalog NumberQuantity
MPB02191431.5500 g
Catalog number MPB02191431.5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
CAS7758-11-4
Recommended StorageRoom Temperature (15-30°C), dessicate.
Physical FormCrystalline Powder
Quantity500 g
Molecular FormulaHK2O4P
View more
Potassium Phosphate occurs in several forms: monobasic, dibasic, and tribasic. Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.