Check Order Status
Potassium Nitrate, (Crystalline/ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Potassium Nitrate, (Crystalline/ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219142801
1 kg
Catalog number
MPB0219142801
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Nitrate
CAS
7757-79-1
Storage Note 1
Not for export
Recommended Storage
Room Temperature
Physical Form
Crystalline
View more