Check Order Status
Potassium Chloride, (Crystalline/ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Potassium Chloride, (Crystalline/ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219142701
1 kg
Catalog number
MPB0219142701
Price (SGD)
208.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
208.00
Each
Add to cart
Specifications
Chemical Name or Material
Potassium Chloride
CAS
7447-40-7
Recommended Storage
Room Temperature
Physical Form
Crystalline
Quantity
1 kg
View more