Check Order Status
Magnesium Chloride Hexahydrate, 99.0-101.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Magnesium Chloride Hexahydrate, 99.0-101.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219142101
1 kg
Catalog number
MPB0219142101
Price (SGD)
307.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
307.00
EA
Add to cart
Specifications
Chemical Name or Material
Magnesium Chloride Hexahydrate
CAS
7791-18-6
Recommended Storage
Room Temperature
Physical Form
Crystalline
Quantity
1 kg
View more