Dimethyl sulfoxide, For ACS analysis, ≥99.9%, MP Biomedicals™
Dimethyl sulfoxide, For ACS analysis, ≥99.9%, MP Biomedicals™
Dimethyl sulfoxide, For ACS analysis, ≥99.9%, MP Biomedicals™
MP Biomedicals
Dimethyl sulfoxide, For ACS analysis, ≥99.9%, MP Biomedicals™
Have Questions?
Catalog NumberQuantity
MPB02191418900.5 L
Catalog number MPB0219141890
Price (SGD)
258.00
Each
Quantity:
0.5 L
Request bulk or custom format
Price (SGD)
258.00
Each
Specifications
Chemical Name or MaterialDimethyl sulfoxide
Name NoteFor ACS analysis
CAS67-68-5
ColorColorless
Density1.104g/cm3 at 20°C
View more