Check Order Status
Dimethyl Sulfoxide, ≥99% (Molecular Biology/ACS Certified), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Dimethyl Sulfoxide, ≥99% (Molecular Biology/ACS Certified), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219141880
100 mL
Catalog number
MPB0219141880
Price (SGD)
146.00
Each
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (SGD)
146.00
Each
Add to cart
Specifications
Chemical Name or Material
Dimethyl Sulfoxide
CAS
67-68-5
Quantity
100 mL
Molecular Formula
C2H6OS
MDL Number
MFCD00002089
View more