Ammonium Chloride, MP Biomedicals™
Ammonium Chloride, MP Biomedicals™
Ammonium Chloride, MP Biomedicals™
Ammonium Chloride, MP Biomedicals™
MP Biomedicals
Ammonium Chloride, MP Biomedicals™
Used in research applications as an ACS reagent. Ammonium Chloride, MP Biomedicals is used in research to lyse human red blood cells, to study basic calcium phosphate crystals in fibroblasts, to isolate proteins from 50S ribosomal subunits of Bacillus ste
Have Questions?
Catalog NumberQuantity
MPB02191406.5500 g
Catalog number MPB02191406.5
Price (SGD)
103.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
103.00
Each
Add to cart
Specifications
CAS12125-02-9
Recommended StorageRoom Temperature, (15-30°C).
Physical FormPowder
Quantity500 g
Molecular FormulaClH4N
View more
Industrial applications include as a flux for coating sheet iron with zinc; tinning; in dry and Leclanch batteries; dyeing, freezing mixtures, electroplating, to clean soldering irons, safety explosives, lustering cotton, tanning; in washing powders; manufacturing of dyes; in cement for iron pipes; for snow treatment (slows melting on ski slopes).