Check Order Status
Sodium Metasilicate, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Sodium Metasilicate, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219138283
250 g
Catalog number
MPB0219138283
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Metasilicate
CAS
13517-24-3
Recommended Storage
Room Temperature
Quantity
250 g
Assay Percent Range
≥97%
View more