Goat Serum: Normal, Sterile
Actual product may vary
MP Biomedicals
Goat Serum: Normal, Sterile
Suitable replacement for FBS on most cell lines and primary cultures for Biomedical Research
Have Questions?
Catalog NumberQuantityProduct Type
MPB0219135680100 mLGoat Serum
Catalog number MPB0219135680
Price (SGD)
132.00
Each
Quantity:
100 mL
Product Type:
Goat Serum
Price (SGD)
132.00
Each
Specifications
Certifications/ComplianceU.S. regulations
ColorRed
SpeciesGoat
Product TypeGoat Serum
Quantity100 mL
SterilitySterile
Sterilization Method0.2 μm filtration
For Use With (Application)Immunohistochemistry
Hemoglobin Concentration<20mg/dL
Mycoplasma DetectedNo
pH6.5 to 8
Unit SizeEach
Contents & Storage
-20°C