Check Order Status
Clindamycin, White Powder, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Clindamycin, White Powder, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219116880
100 mg
Catalog number
MPB0219116880
Price (SGD)
754.00
EA
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (SGD)
754.00
EA
Add to cart
Specifications
Chemical Name or Material
Clindamycin
Type
Clindamycin
Physical Form
Solid
CAS
21462-39-5
Recommended Storage
Store at 4°C.
View more