HISTOPAQUE™ for Lymphocyte Isolation
Product Image
MP Biomedicals
HISTOPAQUE™ for Lymphocyte Isolation
MP Biomedicals™ HISTOPAQUE™ for Lymphocyte Isolation used in isolating lymphocytes from whole blood in areas such as histocompatability testing and in vitro cell-mediated immunity assays
Have Questions?
Catalog NumberQuantity
MPB0219083780100 mL
Catalog number MPB0219083780
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
  • Used for lymphocyte isolation procedure involving a short, low speed centrifugation
  • Easy one-step, rapid, reproducible method for preparation of viable lymphocytes in high yield
  • Method of choice in emergency tissue typing procedures
Specifications
Quantity100 mL
ColorClear
Density1.077 g/mL
SterilitySterile
FormLiquid
Unit SizeEA
Contents & Storage
Store at +4°C. Protect from light.