Check Order Status
Trichloroacetic acid, 99.6%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Trichloroacetic acid, 99.6%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219052190
500 g
Catalog number
MPB0219052190
Price (SGD)
210.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
210.00
EA
Add to cart
Specifications
Chemical Name or Material
Trichloroacetic acid
CAS
76-03-9
Color
White
Assay Percent Range
0.996
Molecular Formula
C2HCl3O2
View more