Check Order Status
Chloramphenicol Crystalline MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Chloramphenicol Crystalline MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219032125
25 g
Catalog number
MPB0219032125
Price (SGD)
113.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
113.00
EA
Add to cart
Specifications
Chemical Name or Material
Chloramphenicol
CAS
56-75-7
Density
1.586 g/mL
Molecular Formula
C11H12Cl2N2O5
Quantity
25 g
View more