Catalase from Aspergillus niger (E.C.1.11.1.6)
Product Image
MP Biomedicals
Catalase from Aspergillus niger (E.C.1.11.1.6)
Lyophilized powder
Have Questions?
Catalog NumberQuantity
MPB02190311011 mU
Catalog number MPB0219031101
Price (SGD)
155.00
EA
Add to cart
Quantity:
1 mU
Price (SGD)
155.00
EA
Add to cart
Activity: 2000U/mg material. Unit Definition: One unit decomposes oneμmole of hydrogen peroxide per minute at 25°C, pH 7.0.
Ref.: Academic Press, New York, Based on the method of Bergmeyer, Methods of Enzymatic Analysis, 438, (1974).
Specifications
Concentration5 mg/mL
ColorBrown to Tan
For Use With (Application)Removal of Peroxides and generation of Oxygen
SourceAspergillus niger
Quantity1 mU
Product TypeCatalase
Molecular Weight (g/mol)≈240000
FormLyophilized Solid
Unit SizeEA
Contents & Storage
-20°C