1x1 image pixel for data collection
2-Mercaptoethanol, ≥98%, MP Biomedicals™
2-Mercaptoethanol, ≥98%, MP Biomedicals™
2-Mercaptoethanol, ≥98%, MP Biomedicals™
2-Mercaptoethanol, ≥98%, MP Biomedicals™
MP Biomedicals
2-Mercaptoethanol, ≥98%, MP Biomedicals™
Have Questions?
Catalog NumberQuantity
MPB0219024290500 mL
Catalog number MPB0219024290
Price (SGD)
154.00
EA
Estimated availability date 21-Sep-2025
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
154.00
EA
Add to cart
Specifications
Chemical Name or Material2-Mercaptoethanol
CAS60-24-2
ColorColorless to Yellow
Density1.12g/cm3 at 20°C
Assay Percent Range≥98%
View more
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link