Check Order Status
Erythromycin, ∽98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Erythromycin, ∽98%, MP Biomedicals™
Erythromycin ∽98%, White Powder
Have Questions?
Catalog Number
Quantity
MPB0219019701
1 g
Catalog number
MPB0219019701
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material
Erythromycin
Type
Erythromycin
Physical Form
Powder
CAS
114-07-8
Recommended Storage
Store at room temperature.
View more
It Used in cell culture applications at approximately 100 mg/liter.
Depending upon the strain of bacteria, erythromycin has been used between 50 and 200 mg/L to control bacterial growth.
Erythromycin resistance can be induced in bacteria.
Erythromycin is a macrolide antibiotic that bind to the 50S ribosomal subunit and inhibit protein synthesis.
This is an antimicrobial substance produced by the growth of certain strains of Streptomyces erythreus.
It is a mixture consisting largely of erythromycin A with lesser amounts of erythromycins B and C.