Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
MP Biomedicals
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Dimethyl Sulfoxide is used to enhance dermal absorption of many chemicals.
Have Questions?
Catalog NumberQuantity
MPB02190186911 L
Catalog number MPB0219018691
Price (SGD)
423.00
Each
Quantity:
1 L
Request bulk or custom format
Price (SGD)
423.00
Each
Specifications
Chemical Name or MaterialDimethyl sulfoxide
CAS67-68-5
ColorColorless
Density1.104 g/cm3 (at 20°C)
Quantity1 L
View more
  • Dimethyl sulfoxide is used to enhance dermal absorption of many chemicals.
  • A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers.
  • Used in antifreeze or hydraulic fluids and as a cryopreservative for cell cultures.
  • Used in the oxidation of thiols and disulfides to sulfonic acids.
  • Used as a PCR cosolvent to help improve yields, especially in long PCR.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.