Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
MP Biomedicals
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Dimethyl Sulfoxide is used to enhance dermal absorption of many chemicals.
Have Questions?
Catalog NumberQuantity
MPB0219018690500 mL
Catalog number MPB0219018690
Price (SGD)
323.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
323.00
EA
Add to cart
Specifications
Chemical Name or MaterialDimethyl Sulfoxide
CAS67-68-5
Density1.104 g/cm3 at 20 °C
Quantity500 mL
Assay Percent Range99+%
View more
  • Dimethyl sulfoxide is used to enhance dermal absorption of many chemicals.
  • A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers.
  • Used in antifreeze or hydraulic fluids and as a cryopreservative for cell cultures.
  • Used in the oxidation of thiols and disulfides to sulfonic acids.
  • Used as a PCR cosolvent to help improve yields, especially in long PCR.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.