1x1 image pixel for data collection
Ampicillin, Anhydrous, 98.24%, MP Biomedicals™
Ampicillin, Anhydrous, 98.24%, MP Biomedicals™
Ampicillin, Anhydrous, 98.24%, MP Biomedicals™
Ampicillin, Anhydrous, 98.24%, MP Biomedicals™
MP Biomedicals
Ampicillin, Anhydrous, 98.24%, MP Biomedicals™
Have Questions?
Catalog NumberQuantity
MPB02190146055 g
Catalog number MPB0219014605
Price (SGD)
378.00
EA
Estimated availability date 21-Sep-2025
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
378.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmpicillin, Anhydrous
TypeAmpicillin, Anhydrous
Name NoteFor HPLC analysis
Physical FormPowder
CAS69-53-4
View more
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link