Check Order Status
Gentamycin Sulfate, USP, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Gentamycin Sulfate, USP, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219005705
5 g
Catalog number
MPB0219005705
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material
Gentamycin Sulfate, USP
Type
Gentamicin Sulfate
Physical Form
Powder
CAS
1405-41-0
Color
White
View more