Dextran Sulfate Sodium Salt, Colitis Grade (36,000 to 50,000), MP Biomedicals™
MP Biomedicals
Dextran Sulfate Sodium Salt, Colitis Grade (36,000 to 50,000), MP Biomedicals™
Induces severe colitis (inflammatory bowel disease) in laboratory animals
Have Questions?
Catalog NumberQuantity
MPB021601102525 g
Catalog number MPB0216011025
Price (SGD)
950.00
Each
Quantity:
25 g
Request bulk or custom format
Price (SGD)
950.00
Each
Chemical Identifiers
CAS9011-18-1
Specifications
Chemical Name or MaterialDextran Sulfate Sodium Salt
ColorWhite
pH5 to 7.11 (1% aqueous solution)
CAS9011-18-1
Physical FormPowder
View more
Induces severe colitis (inflammatory bowel disease) in laboratory animals
Dextran sulfate sodium salt, or DSS, is a synthetic sulfated branched polysaccharide derivative of dextran that has multiple uses in biomedical and clinical research. One important property of DSS is to trigger colitis in mice and rats when administered in drinking water. DSS binds to medium-chain-length fatty acids in the colon and induces intestinal inflammation. Colitis-grade DSS (MW = 36,000 to 50,000) from MP Bio is one of the most widely used products based on peer reviewed scientific publications.
  • Induces severe colitis (inflammatory bowel disease)
  • Highest sulfur content – 19%
  • Highest chirality - +104° of specific rotation
  • Lowest pH – 6.2 at 1% solution
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.