Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Recombinant Proteins
▸
Albumin, Bovine, For IVD, Low Ash and Metals
Actual product may vary
MP Biomedicals
Albumin, Bovine, For IVD, Low Ash and Metals
Prepared by heat shock fractionation
Have Questions?
Change view
Catalog Number
Quantity
MPB0216006980
100 g
MPB0216006910
10 g
2 Options
Catalog number
MPB0216006980
Price (SGD)
651.00
Each
Add to cart
Quantity:
100 g
Price (SGD)
651.00
Each
Add to cart
For IVD applications
Low visible absorbance
Purity: >98%
Ash: <2%
Metals: <20ppm
For best cell culture performance, see alternate Mfr. No. 180577
Specifications
For Use With (Application)
ELISA, Microbial Culture
Molecular Weight (g/mol)
66.338
Name
Bovine Albumin Fraction V
pH Range
6.5 to 7.5
Quantity
100 g
Source
Bovine
Storage Requirements
2° to 8°C
Form
Powder