Check Order Status
2-Methylisothiazolone Hydrochloride, ≥99.9, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
2-Methylisothiazolone Hydrochloride, ≥99.9, MP Biomedicals
Have Questions?
Change view
Quantity:
1 g
500 mg
Catalog number
MPB0215952291
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material
2-Methylisothiazolone Hydrochloride
Type
2-Methylisothiazolone Hydrochloride
Physical Form
Crystalline
CAS
26172-54-3
Recommended Storage
Store at room temperature.
View more