Check Order Status
Xylenes, 99.99%, For ACS analysis, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Xylenes, 99.99%, For ACS analysis, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215869390
500 mL
Catalog number
MPB0215869390
Price (SGD)
91.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
91.00
EA
Add to cart
Specifications
Chemical Name or Material
Xylenes, ACS
Name Note
For ACS analysis
CAS
1330-20-7
Color
Colorless
Density
0.84 g/cm3 at 25°C
View more