Xylene, MP Biomedicals™
Xylene, MP Biomedicals™
Xylene, MP Biomedicals™
Xylene, MP Biomedicals™
MP Biomedicals
Xylene, MP Biomedicals™
Xylene, MP Biomedicals is a solvent used in Giemsa cell staining protocols.
Have Questions?
Catalog NumberQuantity
MPB02158692044 L
Catalog number MPB0215869204
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
CAS1330-20-7
Recommended StorageStore at Room Temperature (15-30°C). Protect from light.
Molecular FormulaC8H10
SynonymXylol, dimethylbenzene, Xylene mixture of isomers
Solubility InformationSoluble in absolute alcohol, ether, many organic solvents; practically in insoluble in water.
View more