Check Order Status
β-Escin, 92 to 95%, MP Biomedicals™
Structure Search
MP Biomedicals
β-Escin, 92 to 95%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215794105
5 g
Catalog number
MPB0215794105
Price (SGD)
580.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
580.00
EA
Add to cart
Specifications
Chemical Name or Material
β-Escin
CAS
11072-93-8
Color
White
Assay Percent Range
92 to 95%
Molecular Formula
C110H172O48
View more