Check Order Status
4',6-Diamidino-2-phenylindole dihydrochloride, >98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
4',6-Diamidino-2-phenylindole dihydrochloride, >98%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215757410
10 mg
Catalog number
MPB0215757410
Price (SGD)
236.00
EA
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (SGD)
236.00
EA
Add to cart
Specifications
Chemical Name or Material
4', 6-Diamidino-2-phenylindole dihydrochloride
CAS
28718-90-3
Color
Yellow to Orange
Assay Percent Range
>98%
Molecular Formula
C16H17Cl2N5
View more