Check Order Status
Heptakis(2,6-di-o-methyl)-β-cyclodextrin, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Heptakis(2,6-di-o-methyl)-β-cyclodextrin, MP Biomedicals™
Heptakis(2,6-di-O-methyl)-β-cyclodextrin is a cholesterol sequestering agent
Have Questions?
Catalog Number
Quantity
MPB0215732005
5 g
Catalog number
MPB0215732005
Price (SGD)
554.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
554.00
EA
Add to cart
Specifications
Chemical Name or Material
Heptakis(2, 6-di-o-methyl)-β-cyclodextrin
CAS
51166-71-3
Color
White
Molecular Formula
C56H98O35
MDL Number
MFCD00011616
View more