Check Order Status
Tamoxifen, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Tamoxifen, MP Biomedicals
Tamoxifen, White Powder
Have Questions?
Catalog Number
Quantity
MPB0215673894
5 g
Catalog number
MPB0215673894
Price (SGD)
1,766.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
1,766.00
EA
Add to cart
Specifications
CAS
10540-29-1
Melting Point
96°C to 100°C
Storage Note 1
Store at +4°C.
Color
White
Loss on Drying
0.0021
View more
Tamoxifen has been used to facilitate the recombination of ect2flox allele in mouse organs
It has also been used to study its effect on lipopolysaccharide (LPS)-induced microglial activation