Check Order Status
Rosin, Gum, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Rosin, Gum, MP Biomedicals™
acid value: 150 to 170
Have Questions?
Catalog Number
Quantity
MPB0215655790
500 g
Catalog number
MPB0215655790
Price (SGD)
205.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
205.00
EA
Add to cart
Specifications
Chemical Name or Material
Rosin, Gum
CAS
8050-09-7
Color
Orange
Recommended Storage
Store at Room Temperature (15 to 30°C).
Density
1.07 - 1.09g/cm
3
at 20°C
View more