Search Thermo Fisher Scientific
| Catalog Number | Quantity |
|---|---|
| MPB0215595725 | 25 g |
Novobiocin inhibits DNA synthesis by inhibiting bacterial DNA gyrase targeting the GyrB subunit of the enzyme involved in energy transduction. It acts as a competitive inhibitor of the ATPase reaction catalysed by GyrB 2. Uses include: