Check Order Status
Indocyanine green, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Indocyanine green, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215502050
50 mg
Catalog number
MPB0215502050
Price (SGD)
256.00
EA
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (SGD)
256.00
EA
Add to cart
Specifications
Chemical Name or Material
Indocyanine green
CAS
3599-32-4
Recommended Storage
Store at Room Temperature (15 to 30°C). Desiccate.
Assay Percent Range
∽ 90%
Molecular Formula
C43H47N2NaO6S2
View more