Yeast Lytic Enzyme
Yeast Lytic Enzyme
Actual product may vary
MP Biomedicals
Yeast Lytic Enzyme
From Arthrobacter luteus
Have Questions?
Catalog NumberQuantity
MPB021535261010 kU
Catalog number MPB0215352610
Price (SGD)
895.00
EA
Add to cart
Quantity:
10 kU
Price (SGD)
895.00
EA
Add to cart
Unit Definition: One unit will produce a ΔA800 of 0.001 per minute at pH 7.5 and 25°C, using a suspension of brewers yeast as a substrate in a 3 mL reaction mixture.
This enzyme catalyzes the lysis of yeast cells and hydrolyzes glucose polymers with β-1,3-linkages (e.g., glucan). It is also useful for preparing yeast spheroplasts.
Zymolyase
Lyophilized powder
Activity: 20,000 units/g
Specifications
Concentration20,000 U/g
ColorTan
pH7.5
For Use With (Application)Digest the cell wall of active Yeast cells
SourceArthrobacter luteus
Quantity10 kU
Product TypeYeast Lytic Enzyme
FormLyophilized Powder
Unit SizeEA
Contents & Storage
0°C