Acid Blue 29, MP Biomedicals™
Acid Blue 29, MP Biomedicals™
Acid Blue 29, MP Biomedicals™
Acid Blue 29, MP Biomedicals™
MP Biomedicals
Acid Blue 29, MP Biomedicals™
Have Questions?
Catalog NumberQuantity
MPB0215259680100 g
Catalog number MPB0215259680
Price (SGD)
232.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
232.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcid Blue 29
CAS5850-35-1
Recommended StorageStore at Room Temperature (15° to 30°C)
EINECS Number227-449-7
Molecular FormulaC22H14N6Na2O9S2
View more
Stains and dyes