Check Order Status
Sodium Sulfide Nonahydrate ACS MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Sodium Sulfide Nonahydrate ACS MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0215258080
100 g
Catalog number
MPB0215258080
Price (SGD)
105.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
105.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium Sulfide
Name Note
Nonahydrate
CAS
1313-84-4
Quantity
100 g
Molecular Formula
H18Na2O9S
View more