Check Order Status
Potassium Bicarbonate, 99.7-100.5%, (ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Potassium Bicarbonate, 99.7-100.5%, (ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0215255780
100 g
Catalog number
MPB0215255780
Price (SGD)
59.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
59.00
EA
Add to cart
Specifications
Chemical Name or Material
Potassium Bicarbonate
CAS
298-14-6
Recommended Storage
Room Temperature
Quantity
100 g
Assay Percent Range
99.7-100.5%
View more