Check Order Status
Ferrous Ammonium Sulfate Hexahydrate, 98.5-101.5%, (ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Ferrous Ammonium Sulfate Hexahydrate, 98.5-101.5%, (ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0215252391
1 kg
Catalog number
MPB0215252391
Price (SGD)
473.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
473.00
EA
Add to cart
Specifications
Chemical Name or Material
Ferrous Ammonium Sulfate Hexahydrate
CAS
7783-85-9
Recommended Storage
Room Temperature
Quantity
1 kg
Assay Percent Range
98.5-101.5%
View more