Check Order Status
Ethylenediamine Tetraacetic Acid Disodium Salt ACS MP Biomedicals
Element Search
Structure Search
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
MP Biomedicals
Ethylenediamine Tetraacetic Acid Disodium Salt ACS MP Biomedicals
Have Questions?
Contact Us
Catalog Number
Quantity
MPB0215252290
500 g
Catalog number
MPB0215252290
Price (SGD)
227.00
EA
Estimated availability date
21-Sep-2025
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
227.00
EA
Add to cart
Specifications
Chemical Name or Material
Ethylenediamine Tetraacetic Acid Disodium Salt
CAS
6381-92-6
Molecular Formula
C10H18N2Na2O10
MDL Number
MFCD00150037,MFCD00003541
Quantity
500 g
View more
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
0 results displayed, search above for a specific certificate
Request a Certificate
Share catalog number, name or link