Check Order Status
Ammonium Molybdate, Tetrahydrate, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Ammonium Molybdate, Tetrahydrate, MP Biomedicals™
Ammonium Molybdate, Tetrahydrate, MP Biomedicals is an analgesic.
Have Questions?
Catalog Number
Quantity
MPB0215249791
1 kg
Catalog number
MPB0215249791
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
CAS
12054-85-2
Recommended Storage
Room Temperature (15–30°C).
Physical Form
Crystalline Powder
Quantity
1 kg
Molecular Formula
(NH4)6Mo7O24.4H2O
View more