Tris, MP Biomedicals™
Tris, MP Biomedicals™
Tris, MP Biomedicals™
Tris, MP Biomedicals™
MP Biomedicals
Tris, MP Biomedicals™
Used as a buffer in a wide variety of biological systems. Tris, MP Biomedicals provides pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis. It can be used as a starting material for polymers, oxazolones (w/ carbox
Have Questions?
Catalog NumberQuantity
MPB02152176.5500 g
Catalog number MPB02152176.5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
CAS77-86-1
Recommended StorageStore at Room Temperature (15-30°C), desiccate.
Physical FormPowder
Quantity500 g
Molecular FormulaC4H11NO3
View more
Tris is relatively non-hygroscopic; however, if needed, Tris (base) can be dried at 100°C for up to 4 hours to remove any water. Neither Tris nor Tris Hydrochloride by themselves provide adequate buffering capacity. Generally the two need to be mixed together to provide a buffer with pH between 7 and 9 to provide adequate buffering. The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0.03 unit per°C decrease in temperature, and decreases 0.03–0.05 unit per ten-fold dilution.