Check Order Status
Spermine, free base, ≥97%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Spermine, free base, ≥97%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215207005
5 g
Catalog number
MPB0215207005
Price (SGD)
541.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
541.00
EA
Add to cart
Specifications
Chemical Name or Material
Spermine
CAS
71-44-3
Color
White
Molecular Formula
C10H26N4
MDL Number
MFCD00008215
View more