Spermidine, free base, 99.5%, MP Biomedicals™
Spermidine, free base, 99.5%, MP Biomedicals™
Spermidine, free base, 99.5%, MP Biomedicals™
Spermidine, free base, 99.5%, MP Biomedicals™
MP Biomedicals
Spermidine, free base, 99.5%, MP Biomedicals™
Have Questions?
Catalog NumberQuantity
MPB02152068011 g
Catalog number MPB0215206801
Price (SGD)
80.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (SGD)
80.00
EA
Add to cart
Specifications
Chemical Name or MaterialSpermidine free base
Name NoteCell Biology and Analysis
CAS124-20-9
ColorColorless
Recommended StorageStore at 4°C, dessicate.
View more