Potassium Phosphate Dibasic, MP Biomedicals™
Potassium Phosphate Dibasic, MP Biomedicals™
Potassium Phosphate Dibasic, MP Biomedicals™
Potassium Phosphate Dibasic, MP Biomedicals™
MP Biomedicals
Potassium Phosphate Dibasic, MP Biomedicals™
Used as a component for a wide variety of media used in the culture of microorganisms and in phosphate buffered saline (PBS). Potassium Phosphate Dibasic, MP Biomedicals is a reagent with high buffering capacity. In addition to helping maintain pH, it sup
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number MPB02151945.5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
CAS7758-11-4
Recommended StorageRoom Temperature(15–30°C), desiccate.
Physical FormPowder
Quantity500 g
Molecular FormulaHK2O4P
View more
Potassium phosphate occurs in several forms: monobasic, dibasic and tribasic. Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.